accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 116272-78-7

IUPAC Name: 1-chloro-3-fluoro-2-[(1E)-2-nitroethenyl]benzene
Synonyms: 1-chloro-3-fluoro-2-[(1E)-2-nitroethenyl]benzene
Molecular Weight (g/mol): 201.58
Molecular Formula: C8H5ClFNO2
InChi Key: JCIAIKBVPVMTBD-SNAWJCMRSA-N
SMILES: [O-][N+](=O)\C=C\C1=C(F)C=CC=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results