accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1163-67-3

IUPAC Name: 3-(phenylcarbamoyl)naphthalen-2-yl acetate
Synonyms: 3-(phenylcarbamoyl)naphthalen-2-yl acetate
Molecular Weight (g/mol): 305.33
Molecular Formula: C19H15NO3
InChi Key: CVJGNNYDVQYHEO-UHFFFAOYSA-N
SMILES: CC(=O)OC1=C(C=C2C=CC=CC2=C1)C(=O)NC1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results