CAS RN 116450-56-7
IUPAC Name:
16-(2-carboxylatophenyl)-3-oxa-9λ⁵,23-diazaheptacyclo[17.7.1.1⁵,⁹.0²,¹⁷.0⁴,¹⁵.0²³,²⁷.0¹³,²⁸]octacosa-1,4,9(28),13,15,17,19(27)-heptaen-9-ylium
Synonyms:
rhodamine 101 rhodamine 640
Molecular Weight (g/mol):
490.6
Molecular Formula:
C32H30N2O3
InChi Key:
MUSLHCJRTRQOSP-UHFFFAOYSA-N
SMILES:
[O-]C(=O)C1=CC=CC=C1C1=C2C=C3CCC[N+]4=C3C(CCC4)=C2OC2=C3CCCN4CCCC(C=C12)=C34
1
–
1
of
1
results
1
Thermo Scientific Chemicals Rhodamine 101 inner salt, 94%, pure
CAS: 116450-56-7 Molecular Formula: C32H30N2O3 Molecular Weight (g/mol): 490.603 MDL Number: MFCD00051383 InChI Key: MUSLHCJRTRQOSP-UHFFFAOYSA-N PubChem CID: 2735129 ChEBI: CHEBI:52314 SMILES: C1CC2=C3C(=C4C(=C2)C(=C5C=C6CCC[N+]7=C6C(=C5O4)CCC7)C8=CC=CC=C8C(=O)[O-])CCCN3C1