accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 116480-60-5

IUPAC Name: 3-(1-methyl-1,2,3,6-tetrahydropyridin-4-yl)-1H-indole-5-carbonitrile
Synonyms: 3-(1-methyl-3,6-dihydro-2H-pyridin-4-yl)-1H-indole-5-carbonitrile
Molecular Weight (g/mol): 237.31
Molecular Formula: C15H15N3
InChi Key: HNOKHIURIJOQKM-UHFFFAOYSA-N
SMILES: CN1CCC(=CC1)C1=CNC2=CC=C(C=C12)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results