accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1165-91-9

IUPAC Name: bis(2,4,6-trichlorophenyl) oxalate
Synonyms: TCPO
Molecular Weight (g/mol): 448.88
Molecular Formula: C14H4Cl6O4
InChi Key: GEVPIWPYWJZSPR-UHFFFAOYSA-N
SMILES: ClC1=CC(Cl)=C(OC(=O)C(=O)OC2=C(Cl)C=C(Cl)C=C2Cl)C(Cl)=C1
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (2)