accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 116529-60-3

IUPAC Name: 2-bromo-3,5-dinitrobenzoic acid
Synonyms: 2-bromo-3,5-dinitrobenzoic acid
Molecular Weight (g/mol): 291.01
Molecular Formula: C7H3BrN2O6
InChi Key: BBKJKUFARRJPRP-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(=CC(=C1Br)[N+]([O-])=O)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results