accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 116530-60-0

IUPAC Name: 5-nitro-2,3-dihydro-1H-inden-2-one
Synonyms: 5-nitro-1,3-dihydroinden-2-one
Molecular Weight (g/mol): 177.16
Molecular Formula: C9H7NO3
InChi Key: VSEBFWRYDORZJI-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC2=C(CC(=O)C2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results