accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 116539-55-0

IUPAC Name: 3-(methylamino)-1-(thiophen-2-yl)propan-1-ol
Synonyms: 3-(methylamino)-1-(thiophen-2-yl)propan-1-ol
Molecular Weight (g/mol): 171.26
Molecular Formula: C8H13NOS
InChi Key: YEJVVFOJMOHFRL-UHFFFAOYNA-N
SMILES: CNCCC(O)C1=CC=CS1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results