accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 116557-46-1

IUPAC Name: 3-bromo-2-methoxyaniline
Synonyms: 3-bromo-2-methoxyaniline
Molecular Weight (g/mol): 202.05
Molecular Formula: C7H8BrNO
InChi Key: ZLODWCIXZJMLJL-UHFFFAOYSA-N
SMILES: COC1=C(N)C=CC=C1Br
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results