accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 116574-71-1

IUPAC Name: tert-butyl (3R)-3-(hydroxymethyl)piperidine-1-carboxylate
Synonyms: tert-butyl (3R)-3-(hydroxymethyl)piperidine-1-carboxylate
Molecular Weight (g/mol): 215.29
Molecular Formula: C11H21NO3
InChi Key: OJCLHERKFHHUTB-SECBINFHSA-N
SMILES: CC(C)(C)OC(=O)N1CCC[C@@H](CO)C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results