CAS RN 1165936-03-7
IUPAC Name:
2-(2-fluoro-4-methylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Synonyms:
2-(2-fluoro-4-methylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Molecular Weight (g/mol):
236.09
Molecular Formula:
C13H18BFO2
InChi Key:
MKWDECBHRFNFGY-UHFFFAOYSA-N
SMILES:
CC1=CC(F)=C(C=C1)B1OC(C)(C)C(C)(C)O1
1
–
1
of
1
results
1
Promotions Available
2-Fluoro-4-methylbenzeneboronic acid pinacol ester, 96%, Thermo Scientific™
2-Fluoro-4-methylbenzeneboronic acid pinacol ester, 96%, Thermo Scientific™
CAS: 1165936-03-7 Molecular Formula: C13H18BFO2 Molecular Weight (g/mol): 236.093 MDL Number: MFCD18779806 InChI Key: MKWDECBHRFNFGY-UHFFFAOYSA-N Synonym: 2-2-fluoro-4-methylphenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane,2-fluoro-4-methylphenylboronic acid pinacol ester,2-fluoro-4-methylbenzeneboronic acid pinacol ester,2-fluoro-4-methylphenyl boronic acid pinacol ester PubChem CID: 69672435 IUPAC Name: 2-(2-fluoro-4-methylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane SMILES: B1(OC(C(O1)(C)C)(C)C)C2=C(C=C(C=C2)C)F