accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 116748-66-4

IUPAC Name: 4'-fluoro-[1,1'-biphenyl]-4-sulfonyl chloride
Synonyms: 4'-fluoro-[1,1'-biphenyl]-4-sulfonyl chloride
Molecular Weight (g/mol): 270.7
Molecular Formula: C12H8ClFO2S
InChi Key: CIDMHDJTWVMBIF-UHFFFAOYSA-N
SMILES: FC1=CC=C(C=C1)C1=CC=C(C=C1)S(Cl)(=O)=O
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)