accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHFFOOF

CAS RN 116751-24-7

IUPAC Name: 2,4,5-trifluoro-3-hydroxybenzoate
Synonyms: 2,4,5-trifluoro-3-hydroxybenzoate
Molecular Weight (g/mol): 191.09
Molecular Formula: C7H2F3O3
InChi Key: YYAFUGSJSHXYNK-UHFFFAOYSA-M
SMILES: OC1=C(F)C(F)=CC(C([O-])=O)=C1F
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results