accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1168135-03-2

IUPAC Name: 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2,1,3-benzothiadiazole
Synonyms: 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2,1,3-benzothiadiazole
Molecular Weight (g/mol): 262.13
Molecular Formula: C12H15BN2O2S
InChi Key: KISHNZJGTMYYKH-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC2=NSN=C2C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results