CAS RN 116855-09-5
IUPAC Name:
1-methyl-1H-pyrazolo[3,4-b]pyridine-3-carboxylic acid
Synonyms:
1-methylpyrazolo[3,4-b]pyridine-3-carboxylic acid
Molecular Weight (g/mol):
177.16
Molecular Formula:
C8H7N3O2
InChi Key:
WEZYUZRHWLFJDN-UHFFFAOYSA-N
SMILES:
CN1N=C(C(O)=O)C2=CC=CN=C12