accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1169402-51-0

BOOH3CCH3H3CH3CNONH2

CAS RN 1169402-51-0

IUPAC Name: 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine-3-carboxamide
Synonyms: 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine-3-carboxamide
Molecular Weight (g/mol): 248.09
Molecular Formula: C12H17BN2O3
InChi Key: NJLZODHYCOZPBJ-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC(=CN=C1)C(N)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results