accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHOOHO

CAS RN 117-10-2

IUPAC Name: 1,8-dihydroxy-9,10-dihydroanthracene-9,10-dione
Synonyms: danthron dantrone [inn-french] danthron (product) dorbanex pastomin 1,8-dihydroanthraquinone danivac chrysazin dioxyanthrachinonum western research brand of danthron
Molecular Weight (g/mol): 240.21
Molecular Formula: C14H8O4
InChi Key: QBPFLULOKWLNNW-UHFFFAOYSA-N
SMILES: OC1=CC=CC2=C1C(=O)C1=C(O)C=CC=C1C2=O
Molecular Weight (g/mol) 
  • (3)
  • (3)
Percent Purity 
  • (1)
  • (5)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (5)
  • (3)
  • (3)
  • (1)
  • (5)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)