accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3ONHINHCH3OIHOOI

CAS RN 117-96-4

IUPAC Name: 3,5-diacetamido-2,4,6-triiodobenzoic acid
Synonyms: diatrizoate meglumine diatrizoate Diat urovist sodium 300 cardiografin 泛影酸 urogranoic acid urotrast acidum diacetylaminotrijodbenzoicum conray 35
Molecular Weight (g/mol): 613.92
Molecular Formula: C11H9I3N2O4
InChi Key: YVPYQUNUQOZFHG-UHFFFAOYSA-N
SMILES: CC(=O)NC1=C(I)C(C(O)=O)=C(I)C(NC(C)=O)=C1I
Physical Form 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)