accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CCH3OOHNOH

CAS RN 117049-14-6

IUPAC Name: tert-butyl N-(2-hydroxy-1-phenylethyl)carbamate
Synonyms: tert-butyl N-(2-hydroxy-1-phenylethyl)carbamate
Molecular Weight (g/mol): 237.3
Molecular Formula: C13H19NO3
InChi Key: IBDIOGYTZBKRGI-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)NC(CO)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)
Grade 
  • (2)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)