accessibility menu, dialog, popup

UserName

Viewing profile as user:
BONH2HO

CAS RN 117098-94-9

IUPAC Name: 6-amino-1,3-dihydro-2,1-benzoxaborol-1-ol
Synonyms: 6-amino-3H-2,1-benzoxaborol-1-ol
Molecular Weight (g/mol): 148.96
Molecular Formula: C7H8BNO2
InChi Key: USCKQKQTFCBECF-UHFFFAOYSA-N
SMILES: NC1=CC2=C(COB2O)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results