accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 117106-21-5

CH3CH3H3COOHNHNOOOHO

CAS RN 117106-21-5

IUPAC Name: 2-{[(tert-butoxy)carbonyl]amino}-4-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)butanoic acid
Synonyms: 2-[(tert-butoxycarbonyl)amino]-4-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}butanoic acid
Molecular Weight (g/mol): 440.5
Molecular Formula: C24H28N2O6
InChi Key: MJZDTTZGQUEOBL-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)NC(CCNC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)