CAS RN 1171920-23-2
CAS RN 1171920-23-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Promotions Available
Sigma Aldrich 6-Bromo-2-((tert-butyldimethylsilyloxy)methyl)furo[3,2-b]pyridine
Sigma Aldrich 6-Bromo-2-((tert-butyldimethylsilyloxy)methyl)furo[3,2-b]pyridine
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.