accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OPOH3NOH3C

CAS RN 117196-73-3

IUPAC Name: diethyl (azaniumylmethyl)phosphonate
Synonyms: diethyl ammoniomethylphosphonate
Molecular Weight (g/mol): 168.15
Molecular Formula: C5H15NO3P
InChi Key: UIBCDEFKKLRXHR-UHFFFAOYSA-O
SMILES: CCOP(=O)(C[NH3+])OCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results