accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1172-76-5

H3CSiSiCH3

CAS RN 1172-76-5

IUPAC Name: 1,2-dimethyl-1,1,2,2-tetraphenyldisilane
Synonyms: 1,2-dimethyl-1,1,2,2-tetraphenyldisilane
Molecular Weight (g/mol): 394.66
Molecular Formula: C26H26Si2
InChi Key: JNZRJYXUMDPPRK-UHFFFAOYSA-N
SMILES: C[Si](C1=CC=CC=C1)(C1=CC=CC=C1)[Si](C)(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)