accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNNNFPFFFFFFPFFFFFFPFFFFFFPFFFFF

CAS RN 117271-77-9

IUPAC Name: 5λ⁵,12λ⁵,19λ⁵,26λ⁵-tetraazaheptacyclo[24.2.2.2²,⁵.2⁷,¹⁰.2¹²,¹⁵.2¹⁶,¹⁹.2²¹,²⁴]tetraconta-1(28),2,4,7,9,12,14,16,18,21,23,26,29,31,33,35,37,39-octadecaene-5,12,19,26-tetrakis(ylium); tetrakis(hexafluoro-λ⁵-phosphanuide)
Synonyms: cyclobis(paraquat-p-phenylene) tetrahexafluorophosphate
Molecular Weight (g/mol): 1100.54
Molecular Formula: C36H32F24N4P4
InChi Key: ORRBEFRZPYDCAJ-UHFFFAOYSA-N
SMILES: F[P-](F)(F)(F)(F)F.F[P-](F)(F)(F)(F)F.F[P-](F)(F)(F)(F)F.F[P-](F)(F)(F)(F)F.C1C2=CC=C(C[N+]3=CC=C(C=C3)C3=CC=[N+](CC4=CC=C(C[N+]5=CC=C(C=C5)C5=CC=[N+]1C=C5)C=C4)C=C3)C=C2
Grade 
  • (1)
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results