accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNBrBr

CAS RN 1173071-58-3

IUPAC Name: 2,7-dibromo-9-heptyl-9H-carbazole
Synonyms: 2,7-dibromo-9-heptylcarbazole
Molecular Weight (g/mol): 423.19
Molecular Formula: C19H21Br2N
InChi Key: MLLYLIXYBBORGG-UHFFFAOYSA-N
SMILES: CCCCCCCN1C2=C(C=CC(Br)=C2)C2=C1C=C(Br)C=C2
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results