CAS RN 117322-30-2
IUPAC Name:
1-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)cyclopentane-1-carboxylate
Synonyms:
1-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}cyclopentane-1-carboxylate
Molecular Weight (g/mol):
350.4
Molecular Formula:
C21H20NO4
InChi Key:
IECZEINPZOFWNU-UHFFFAOYSA-M
SMILES:
[O-]C(=O)C1(CCCC1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
1
–
1
of
1
results
1
1-[[(9H-Fluoren-9-ylmethoxy)carbonyl]amino]cyclopentanecarboxylic Acid 95.0+%, TCI America™
CAS: 117322-30-2 Molecular Formula: C21H21NO4 Molecular Weight (g/mol): 351.402 MDL Number: MFCD01074696 InChI Key: IECZEINPZOFWNU-UHFFFAOYSA-N Synonym: 1-(Fmoc-amino)cyclopentanecarboxylic Acid, Fmoc-cycloleucine PubChem CID: 978345 IUPAC Name: 1-(9H-fluoren-9-ylmethoxycarbonylamino)cyclopentane-1-carboxylic acid SMILES: C1CCC(C1)(C(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24