CAS RN 117324-58-0
IUPAC Name:
methyl 2-amino-5-(trifluoromethyl)benzoate
Synonyms:
methyl 2-amino-5-(trifluoromethyl)benzoate
Molecular Weight (g/mol):
219.16
Molecular Formula:
C9H8F3NO2
InChi Key:
QGFUDNJZHZNPCS-UHFFFAOYSA-N
SMILES:
COC(=O)C1=C(N)C=CC(=C1)C(F)(F)F
1
–
1
of
1
results
1
Promotions Available
Methyl 2-amino-5-(trifluoromethyl)benzoate, 98%
Methyl 2-amino-5-(trifluoromethyl)benzoate, 98%
CAS: 117324-58-0 Molecular Formula: C9H8F3NO2 Molecular Weight (g/mol): 219.163 MDL Number: MFCD08234902 InChI Key: QGFUDNJZHZNPCS-UHFFFAOYSA-N PubChem CID: 14233808 IUPAC Name: methyl 2-amino-5-(trifluoromethyl)benzoate SMILES: COC(=O)C1=C(C=CC(=C1)C(F)(F)F)N