accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 117344-32-8

IUPAC Name: 2-(4-{9-[4-(2-hydroxyethoxy)phenyl]-9H-fluoren-9-yl}phenoxy)ethan-1-ol
Synonyms: 2-(4-{9-[4-(2-hydroxyethoxy)phenyl]fluoren-9-yl}phenoxy)ethanol
Molecular Weight (g/mol): 438.52
Molecular Formula: C29H26O4
InChi Key: NQXNYVAALXGLQT-UHFFFAOYSA-N
SMILES: OCCOC1=CC=C(C=C1)C1(C2=CC=CC=C2C2=CC=CC=C12)C1=CC=C(OCCO)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results