accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3H3COOOOH3CCH3H3COHHO

CAS RN 117384-45-9

IUPAC Name: 1,4-di-tert-butyl 2,3-dihydroxybutanedioate
Synonyms: 1,4-di-tert-butyl 2,3-dihydroxybutanedioate
Molecular Weight (g/mol): 262.3
Molecular Formula: C12H22O6
InChi Key: ITWOKJQQGHCDBL-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)C(O)C(O)C(=O)OC(C)(C)C
Percent Purity 
  • (1)
  • (3)
Physical Form 
  • (1)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)