CAS RN 1174006-47-3
IUPAC Name:
1-(4-{9-[4-(pyren-1-yl)phenyl]-9H-fluoren-9-yl}phenyl)pyrene
Synonyms:
1-(4-{9-[4-(pyren-1-yl)phenyl]fluoren-9-yl}phenyl)pyrene
Molecular Weight (g/mol):
718.9
Molecular Formula:
C57H34
InChi Key:
WHLKCPQPGTUZBU-UHFFFAOYSA-N
SMILES:
C1=CC=C2C(=C1)C1=CC=CC=C1C2(C1=CC=C(C=C1)C1=CC=C2C=CC3=CC=CC4=CC=C1C2=C34)C1=CC=C(C=C1)C1=CC=C2C=CC3=CC=CC4=CC=C1C2=C34
1
–
1
of
1
results
1
9,9-Bis[4-(1-pyrenyl)phenyl]fluorene 96.0+%, TCI America™
CAS: 1174006-47-3 Molecular Formula: C57H34 Molecular Weight (g/mol): 718.899 MDL Number: MFCD12022454 InChI Key: WHLKCPQPGTUZBU-UHFFFAOYSA-N PubChem CID: 101502333 IUPAC Name: 1-[4-[9-(4-pyren-1-ylphenyl)fluoren-9-yl]phenyl]pyrene SMILES: C1=CC=C2C(=C1)C3=CC=CC=C3C2(C4=CC=C(C=C4)C5=C6C=CC7=CC=CC8=C7C6=C(C=C8)C=C5)C9=CC=C(C=C9)C1=C2C=CC3=CC=CC4=C3C2=C(C=C4)C=C1