accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNBrSOOCl

CAS RN 1174064-62-0

IUPAC Name: 4-(4-bromo-1H-pyrazol-1-yl)benzene-1-sulfonyl chloride
Synonyms: 4-(4-bromopyrazol-1-yl)benzenesulfonyl chloride
Molecular Weight (g/mol): 321.57
Molecular Formula: C9H6BrClN2O2S
InChi Key: LRYXAJMNOSSJDD-UHFFFAOYSA-N
SMILES: ClS(=O)(=O)C1=CC=C(C=C1)N1C=C(Br)C=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results