accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 117666-97-4

IUPAC Name: 3-(4-benzoylphenyl)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)propanoic acid
Synonyms: 3-(4-benzoylphenyl)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}propanoic acid
Molecular Weight (g/mol): 491.54
Molecular Formula: C31H25NO5
InChi Key: SYOBJKCXNRQOGA-UHFFFAOYNA-N
SMILES: OC(=O)C(CC1=CC=C(C=C1)C(=O)C1=CC=CC=C1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)