accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOBrCl

CAS RN 117738-74-6

IUPAC Name: methyl 4-bromo-3-chlorobenzoate
Synonyms: methyl 4-bromo-3-chlorobenzoate
Molecular Weight (g/mol): 249.49
Molecular Formula: C8H6BrClO2
InChi Key: BPJMNAWSHVKGHE-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC(Cl)=C(Br)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results