accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 117766-40-2

H3COOH3C

CAS RN 117766-40-2

IUPAC Name: 9,9-bis(4-methoxyphenyl)-9H-fluorene
Synonyms: 9,9-bis(4-methoxyphenyl)fluorene
Molecular Weight (g/mol): 378.47
Molecular Formula: C27H22O2
InChi Key: BYDIWLYAWBNCAQ-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1)C1(C2=CC=CC=C2C2=CC=CC=C12)C1=CC=C(OC)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)