CAS RN 1177865-17-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ Rac1 Inhibitor
Rac1 Inhibitor, CAS 1177865-17-6, is a cell-permeable, reversible inhibitor of Rac1 GDP/GTP exchange. Interferes with the interaction between Rac1 and Rac-specific GEFs Trio and Tiam1 (IC50 ≈50 μM).