accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3C(R)O(S)HNOHOOO

CAS RN 117872-75-0

IUPAC Name: (2S,3R)-3-(benzyloxy)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)butanoic acid
Synonyms: (2S,3R)-3-(benzyloxy)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}butanoic acid|FMOC-THR(BZL)-OH
Molecular Weight (g/mol): 431.49
Molecular Formula: C26H25NO5
InChi Key: UCDMMWCWPVCHLL-OSPHWJPCSA-N
SMILES: C[C@@H](OCC1=CC=CC=C1)[C@H](NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results