accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOOO

CAS RN 117953-13-6

IUPAC Name: 5-(1,3-dioxolan-2-yl)furan-2-carbaldehyde
Synonyms: 5-(1,3-dioxolan-2-yl)furan-2-carbaldehyde
Molecular Weight (g/mol): 168.15
Molecular Formula: C8H8O4
InChi Key: XTPZCGOSUHCSQZ-UHFFFAOYSA-N
SMILES: O=CC1=CC=C(O1)C1OCCO1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results