accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 118-41-2

IUPAC Name: 3,4,5-trimethoxybenzoic acid
Synonyms: 3,4,5-trimethoxybenzoic acid tri-O-methylgallic acid trimethylgallic acid eudesmic acid gallic acid trimethyl ether
Molecular Weight (g/mol): 212.2
Molecular Formula: C10H12O5
InChi Key: SJSOFNCYXJUNBT-UHFFFAOYSA-N
SMILES: COC1=CC(=CC(OC)=C1OC)C(O)=O
Boiling Point 
  • (2)
  • (1)
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (2)
  • (3)
  • (1)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (2)
  • (5)
  • (2)
  • (1)
  • (3)
  • (2)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (2)
  • (3)