accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOCH3CH3CH3CH3CH3

CAS RN 1182-66-7

IUPAC Name: 9a,11a-dimethyl-1-(6-methylheptan-2-yl)-1H,2H,3H,3aH,3bH,4H,6H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-7-yl nonanoate
Synonyms: cholesteryl nonanoate
Molecular Weight (g/mol): 526.89
Molecular Formula: C36H62O2
InChi Key: WCLNGBQPTVENHV-UHFFFAOYNA-N
SMILES: CCCCCCCCC(=O)OC1CCC2(C)C3CCC4(C)C(CCC4C3CC=C2C1)C(C)CCCC(C)C
Quantity 
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (1)
  • (2)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)