accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 118289-55-7

IUPAC Name: 6-chloro-5-(2-chloroethyl)-2,3-dihydro-1H-indol-2-one
Synonyms: 6-chloro-5-(2-chloroethyl)-1,3-dihydroindol-2-one
Molecular Weight (g/mol): 230.09
Molecular Formula: C10H9Cl2NO
InChi Key: ZTQQXEPZEYIVDK-UHFFFAOYSA-N
SMILES: ClCCC1=C(Cl)C=C2NC(=O)CC2=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results