accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COCH3CH3NHONH2NNH2HNOH3CHNOH3CNCH3OHNOCH3OHOHOO(E)(E)(E)

CAS RN 118399-22-7

IUPAC Name: (2E)-9-{3-[(diaminomethylidene)amino]propyl}-2-ethylidene-12-[(1E,3E)-6-methoxy-3,5-dimethyl-7-phenylhepta-1,3-dien-1-yl]-1,6,13-trimethyl-3,7,10,14,19-pentaoxo-1,4,8,11,15-pentaazacyclononadecane-5,16-dicarboxylic acid
Synonyms: nodularin
Molecular Weight (g/mol): 824.98
Molecular Formula: C41H60N8O10
InChi Key: IXBQSRWSVIBXNC-ANPQUZCZNA-N
SMILES: COC(CC1=CC=CC=C1)C(C)\C=C(/C)\C=C\C1NC(=O)C(CCCN=C(N)N)NC(=O)C(C)C(NC(=O)\C(=C/C)N(C)C(=O)CCC(NC(=O)C1C)C(O)=O)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)