accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3HOCH3CH3HOCH3CH3HOCH3CH3HOSn

CAS RN 1184-61-8

IUPAC Name: tetrakis(propan-2-ol) tin
Synonyms: tetrakis(isopropyl alcohol) tin
Molecular Weight (g/mol): 359.09
Molecular Formula: C12H32O4Sn
InChi Key: VUDAJANAMZGVOP-UHFFFAOYSA-N
SMILES: [Sn].CC(C)O.CC(C)O.CC(C)O.CC(C)O
Percent Purity 
  • (4)
  • (4)
Quantity 
  • (2)
  • (2)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (4)
  • (8)
  • (2)
  • (2)
  • (2)
  • (2)