CAS RN 1184023-09-3
IUPAC Name:
2-(4-bromo-1H-pyrazol-1-yl)benzonitrile
Synonyms:
2-(4-bromopyrazol-1-yl)benzonitrile
Molecular Weight (g/mol):
248.08
Molecular Formula:
C10H6BrN3
InChi Key:
RPTLEHPSCLVUDJ-UHFFFAOYSA-N
SMILES:
BrC1=CN(N=C1)C1=CC=CC=C1C#N
1
–
1
of
1
results
1
2-(4-Bromo-1-pyrazolyl)benzonitrile, 98%, Thermo Scientific™
CAS: 1184023-09-3 Molecular Formula: C10H6BrN3 Molecular Weight (g/mol): 248.083 InChI Key: RPTLEHPSCLVUDJ-UHFFFAOYSA-N Synonym: 2-4-bromopyrazol-1-yl benzonitrile,2-4-bromo-1h-pyrazol-1-yl benzonitrile,2-4-bromo-1-pyrazolyl benzonitrile PubChem CID: 61035135 IUPAC Name: 2-(4-bromopyrazol-1-yl)benzonitrile SMILES: C1=CC=C(C(=C1)C#N)N2C=C(C=N2)Br