accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNNNH2

CAS RN 118430-74-3

IUPAC Name: 3-cyclopropyl-1-methyl-1H-pyrazol-5-amine
Synonyms: 5-cyclopropyl-2-methylpyrazol-3-amine
Molecular Weight (g/mol): 137.19
Molecular Formula: C7H11N3
InChi Key: YWHWPIRLFHZSFS-UHFFFAOYSA-N
SMILES: CN1N=C(C=C1N)C1CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results