accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 118465-49-9

IUPAC Name: methyl 6H-thieno[2,3-b]pyrrole-5-carboxylate
Synonyms: methyl 6H-thieno[2,3-b]pyrrole-5-carboxylate
Molecular Weight (g/mol): 181.21
Molecular Formula: C8H7NO2S
InChi Key: YBIAQTPXONLBJG-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC2=C(N1)SC=C2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results