accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 118486-97-8

IUPAC Name: 1-methyl-2-(tributylstannyl)-1H-pyrrole
Synonyms: 1-methyl-2-(tributylstannyl)pyrrole
Molecular Weight (g/mol): 370.17
Molecular Formula: C17H33NSn
InChi Key: DINAKCGOEKXDTP-UHFFFAOYSA-N
SMILES: CCCC[Sn](CCCC)(CCCC)C1=CC=CN1C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results