accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1185-39-3

CH3H3CCH3OHO

CAS RN 1185-39-3

IUPAC Name: 2,2-dimethylpentanoic acid
Synonyms: 2,2-dimethylvaleric acid
Molecular Weight (g/mol): 130.19
Molecular Formula: C7H14O2
InChi Key: ZRYCZAWRXHAAPZ-UHFFFAOYSA-N
SMILES: CCCC(C)(C)C(O)=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results