accessibility menu, dialog, popup

UserName

Viewing profile as user:
HNONNHCl

CAS RN 1185136-22-4

IUPAC Name: 6-(4-{[1,1'-biphenyl]-2-yl}piperazin-1-yl)-N-(1,2,3,4-tetrahydronaphthalen-1-yl)hexanamide hydrochloride
Synonyms: 6-(4-{[1,1'-biphenyl]-2-yl}piperazin-1-yl)-N-(1,2,3,4-tetrahydronaphthalen-1-yl)hexanamide hydrochloride
Molecular Weight (g/mol): 518.14
Molecular Formula: C32H40ClN3O
InChi Key: XNDUOTNYUWDFKR-UHFFFAOYNA-N
SMILES: Cl.O=C(CCCCCN1CCN(CC1)C1=CC=CC=C1C1=CC=CC=C1)NC1CCCC2=CC=CC=C12
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results