CAS RN 1185310-49-9
IUPAC Name:
6-bromo-4-methyl-3-nitroquinoline
Synonyms:
6-bromo-4-methyl-3-nitroquinoline
Molecular Weight (g/mol):
267.08
Molecular Formula:
C10H7BrN2O2
InChi Key:
MYJZQOHLSJFXDS-UHFFFAOYSA-N
SMILES:
CC1=C(C=NC2=CC=C(Br)C=C12)[N+]([O-])=O
1
–
1
of
1
results
1
Promotions Available
6-Bromo-4-methyl-3-nitroquinoline, 96%, Thermo Scientific Chemicals
6-Bromo-4-methyl-3-nitroquinoline, 96%, Thermo Scientific Chemicals
CAS: 1185310-49-9 Molecular Formula: C10H7BrN2O2 Molecular Weight (g/mol): 267.082 MDL Number: MFCD12761249 InChI Key: MYJZQOHLSJFXDS-UHFFFAOYSA-N PubChem CID: 45789224 IUPAC Name: 6-bromo-4-methyl-3-nitroquinoline SMILES: CC1=C2C=C(C=CC2=NC=C1[N+](=O)[O-])Br